ENA245486953 Enamine

methyl 5-(chlorosulfonyl)-2-fluorobenzoate

  • MDL number MFCD11650665
  • Cas Number : 1094460-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54846
Availability details for
Quantity
  • Availability
  • Add to Cart