ENA245486956 Enamine

methyl 2-[5-(chlorosulfonyl)thiophen-2-yl]acetate

  • MDL number MFCD12169467
  • Cas Number : 933730-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54850

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54850
Availability details for
Quantity
  • Availability
  • Add to Cart