ENA245486957 Enamine

methyl 3-(chlorosulfonyl)-4,5-dimethylbenzoate

  • MDL number MFCD12170853
  • Cas Number : 1156634-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54852
Availability details for
Quantity
  • Availability
  • Add to Cart