ENA245486960 Enamine

1-(2-phenylacetyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00451541
  • Cas Number : 2752-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54856
Availability details for
Quantity
  • Availability
  • Add to Cart