ENA245486961 Enamine

3-(cyclohexyloxy)propanimidamide hydrochloride

  • MDL number MFCD13196145
  • Cas Number : 1210660-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54857
Availability details for
Quantity
  • Availability
  • Add to Cart