ENA245486963 Enamine

3-(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)propanoic acid

  • MDL number MFCD09880468
  • Cas Number : 26366-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54859
Availability details for
Quantity
  • Availability
  • Add to Cart