ENA245486965 Enamine

N-(3-amino-2-methylphenyl)butanamide

  • MDL number MFCD09805380
  • Cas Number : 946725-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54861

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54861
Availability details for
Quantity
  • Availability
  • Add to Cart