ENA245486967 Enamine

methyl 2-(4-bromophenoxy)acetate

  • MDL number MFCD01124700
  • Cas Number : 4841-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54863
Availability details for
Quantity
  • Availability
  • Add to Cart