ENA245486970 Enamine

4-amino-1-(4-phenoxyphenyl)pyrrolidin-2-one

  • MDL number MFCD12678737
  • Cas Number : 1155622-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54866

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54866
Availability details for
Quantity
  • Availability
  • Add to Cart