ENA245486973 Enamine

1-amino-3-(1,3-dioxaindan-5-yloxy)propan-2-ol

  • MDL number MFCD04215050
  • Cas Number : 258853-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54870
Availability details for
Quantity
  • Availability
  • Add to Cart