ENA245486983 Enamine

{1-[3-(trifluoromethyl)phenyl]cyclopentyl}methanamine

  • MDL number MFCD10690922
  • Cas Number : 1152568-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54881
Availability details for
Quantity
  • Availability
  • Add to Cart