ENA245486991 Enamine

2-[(2,6-dimethoxyphenoxy)methyl]oxirane

  • MDL number MFCD09740100
  • Cas Number : 5435-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54889
Availability details for
Quantity
  • Availability
  • Add to Cart