ENA245486998 Enamine

4-[(piperidin-1-yl)methyl]piperidine dihydrochloride

  • MDL number MFCD12407022
  • Cas Number : 32832-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54899
Availability details for
Quantity
  • Availability
  • Add to Cart