ENA245487002 Enamine

5-bromo-N-methylthiophene-2-sulfonamide

  • MDL number MFCD03856196
  • Cas Number : 81597-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54903

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54903
Availability details for
Quantity
  • Availability
  • Add to Cart