ENA245487010 Enamine

4-(3-chloro-4-fluorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD03424575
  • Cas Number : 145029-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53844
Availability details for
Quantity
  • Availability
  • Add to Cart