ENA245487013 Enamine

1-ethoxypropan-2-amine hydrochloride

  • MDL number MFCD09879167
  • Cas Number : 1185304-14-6
  • Molecular Weight : 139.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54916
Availability details for
Quantity
  • Availability
  • Add to Cart