ENA245487015 Enamine

methyl[(2-methylfuran-3-yl)methyl]amine

  • MDL number MFCD09702377
  • Cas Number : 916766-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54919

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54919
Availability details for
Quantity
  • Availability
  • Add to Cart