ENA245487022 Enamine

2-ethoxy-2-phenylacetic acid

  • MDL number MFCD09948905
  • Cas Number : 33224-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54926

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54926
Availability details for
Quantity
  • Availability
  • Add to Cart