ENA245487023 Enamine

2-methoxyethyl N-(3-aminophenyl)carbamate

  • MDL number MFCD11638281
  • Cas Number : 1095601-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54927
Availability details for
Quantity
  • Availability
  • Add to Cart