ENA245487028 Enamine

2-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-phenylacetamide

  • MDL number MFCD09041764
  • Cas Number : 926198-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54933
Availability details for
Quantity
  • Availability
  • Add to Cart