ENA245487038 Enamine

3-[(2-methoxy-5-methylphenyl)amino]propanoic acid

  • MDL number MFCD11186115
  • Cas Number : 23363-80-6
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54953
Availability details for
Quantity
  • Availability
  • Add to Cart