ENA245487039 Enamine

1-amino-3-[3-(trifluoromethyl)phenoxy]propan-2-ol

  • MDL number MFCD11131172
  • Cas Number : 4698-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54954
Availability details for
Quantity
  • Availability
  • Add to Cart