ENA245487043 Enamine

3-(5-phenyl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD09045564
  • Cas Number : 926240-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-54959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54959
Availability details for
Quantity
  • Availability
  • Add to Cart