ENA245487046 Enamine

1-(5-methyl-1,2-oxazol-3-yl)methanamine

  • MDL number MFCD02681995
  • Cas Number : 154016-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54966

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-54966
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart