ENA245487048 Enamine

2-methyl-6-phenylmorpholine

  • MDL number MFCD11652903
  • Cas Number : 59630-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54968

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54968
Availability details for
Quantity
  • Availability
  • Add to Cart