ENA245487050 Enamine

2-(3,4-dimethylphenyl)oxirane

  • MDL number MFCD01741454
  • Cas Number : 1855-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 70
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54970
Availability details for
Quantity
  • Availability
  • Add to Cart