ENA245487056 Enamine

(1R)-1-(3,4-difluorophenyl)ethan-1-ol

  • MDL number MFCD09863750
  • Cas Number : 126534-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54981
Availability details for
Quantity
  • Availability
  • Add to Cart