ENA245487058 Enamine

3-(2,6-dimethylmorpholin-4-yl)propanimidamide dihydrochloride

  • MDL number MFCD13196161
  • Cas Number : 1208701-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54988
Availability details for
Quantity
  • Availability
  • Add to Cart