ENA245487063 Enamine

2-[(2-cyanophenyl)sulfanyl]acetic acid

  • MDL number MFCD11155589
  • Cas Number : 98589-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54999
Availability details for
Quantity
  • Availability
  • Add to Cart