ENA245487069 Enamine

1-(2-chlorophenyl)butane-1,3-dione

  • MDL number MFCD07772928
  • Cas Number : 56464-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55006
Availability details for
Quantity
  • Availability
  • Add to Cart