ENA245487070 Enamine

4-(pentyloxy)benzene-1-sulfonyl chloride

  • MDL number MFCD01961265
  • Cas Number : 58076-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55007
Availability details for
Quantity
  • Availability
  • Add to Cart