ENA245487075 Enamine

2-[(trifluoromethyl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD09040625
  • Cas Number : 929971-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55020

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55020
Availability details for
Quantity
  • Availability
  • Add to Cart