ENA245487084 Enamine

6-ethyl-2-phenyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD12137535
  • Cas Number : 83501-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55032
Availability details for
Quantity
  • Availability
  • Add to Cart