ENA245487087 Enamine

3-(2,2-dimethylpropyl)-[1,2]oxazolo[5,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09738328
  • Cas Number : 954277-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55035
Availability details for
Quantity
  • Availability
  • Add to Cart