ENA245487088 Enamine

3-propyl-[1,2]oxazolo[5,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09731156
  • Cas Number : 953885-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55036
Availability details for
Quantity
  • Availability
  • Add to Cart