ENA245487090 Enamine

5-[(2-aminoethyl)amino]-4-chloro-2,3-dihydropyridazin-3-one

  • MDL number MFCD09041602
  • Cas Number : 83394-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55038
Availability details for
Quantity
  • Availability
  • Add to Cart