ENA245487097 Enamine

1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)ethan-1-one

  • MDL number MFCD00514548
  • Cas Number : 1210-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55054

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55054
Availability details for
Quantity
  • Availability
  • Add to Cart