ENA245487100 Enamine

2,2,2-trifluoroethyl N-(pyridin-2-yl)carbamate

  • MDL number MFCD11099824
  • Cas Number : 1087788-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55059
Availability details for
Quantity
  • Availability
  • Add to Cart