ENA245487103 Enamine

2-(phenylmethane)sulfonylethan-1-amine

  • MDL number MFCD09939410
  • Cas Number : 46207-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55062
Availability details for
Quantity
  • Availability
  • Add to Cart