ENA245487107 Enamine

2-methyl-3-oxo-3-(pyridin-3-yl)propanenitrile

  • MDL number MFCD11208744
  • Cas Number : 1071089-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55067
Availability details for
Quantity
  • Availability
  • Add to Cart