ENA245487110 Enamine

4-(2,3-dihydro-1,4-benzodioxin-6-yl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD11632168
  • Cas Number : 925006-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55070
Availability details for
Quantity
  • Availability
  • Add to Cart