ENA245487114 Enamine

(3-chlorophenyl)methanethiol

  • MDL number MFCD00068526
  • Cas Number : 25697-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55076

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-55076
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart