ENA245487126 Enamine

1-(2-aminoacetyl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD13196175
  • Cas Number : 1210438-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55093
Availability details for
Quantity
  • Availability
  • Add to Cart