ENA245487129 Enamine

(1-{[(3,4-dichlorophenyl)methyl]amino}-3-methylbutan-2-yl)dimethylamine

  • MDL number MFCD11117609
  • Cas Number : 1036614-61-5
  • Molecular Weight : 289.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N~1~-(3,4-dichlorobenzyl)-N~2~,N~2~,3-trimethyl-1,2-butanediamine
Inchi Code 1S/C14H22Cl2N2/c1-10(2)14(18(3)4)9-17-8-11-5-6-12(15)13(16)7-11/h5-7,10,14,17H,8-9H2,1-4H3
Inchi Key IWSCGDKRPZIJCK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55096

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-55096
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart