ENA245487132 Enamine

1-(3-methylpiperidin-1-yl)propan-2-amine

  • MDL number MFCD09729926
  • Cas Number : 86559-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53856
Availability details for
Quantity
  • Availability
  • Add to Cart