ENA245487134 Enamine

N-benzyl-5-methoxy-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD06604089
  • Cas Number : 233605-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55121
Availability details for
Quantity
  • Availability
  • Add to Cart