ENA245487135 Enamine

2-(3-aminophenyl)-2-azaspiro[4.4]nonane-1,3-dione

  • MDL number MFCD11979596
  • Cas Number : 1154571-03-5
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55123
Availability details for
Quantity
  • Availability
  • Add to Cart