ENA245487137 Enamine

[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD00226396
  • Cas Number : 13223-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55128
Availability details for
Quantity
  • Availability
  • Add to Cart