ENA245487141 Enamine

4-(3,5-dimethyl-1H-pyrazol-1-yl)-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD12557247
  • Cas Number : 1152574-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55135
Availability details for
Quantity
  • Availability
  • Add to Cart