ENA245487142 Enamine

methyl 3-amino-5-(3,5-dichlorophenyl)thiophene-2-carboxylate

  • MDL number MFCD11104878
  • Cas Number : 1152576-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55136
Availability details for
Quantity
  • Availability
  • Add to Cart